Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Irish Moss Carrageenan, MP Biomedicals™

Catalog No. 0210488691 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
02-104-88691 1 kg
ICN10488690 500 g
ICN10488680 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 02-104-88691 Supplier MP Biomedicals, Inc Supplier No. 0210488691
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Irish Moss Carrageenan solution is used to suppress immune response in vivo and in vitro via mechanisms.

  • Carrageenans are used to suppress immune response in vivo and in vitro via mechanisms believed to involve selective cytopathic effect on macrophages.
  • As ι-, κ-, and λ-carrageenans have varying degress of sulfation, they may be studied comparatively for differences in protective, antiviral, and immunosuppression activities.
  • Carrageenan may be used in the development of biomaterials such as bone graft substitutes, for entrapping vessels such as biobeads, and as encapsulation vehicles for drug delivery.
  • Carrageenan can be used to develop hydrogels.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9000-07-1
MDL Number MFCD00081480
Synonym Carrageenan, Gelatin

Specifications

Color Tan
Quantity 1 kg
Physical Form Powder
Chemical Name or Material Irish Moss (Carrageenan), Predominantly Kappa-Carrageenan with lesser amount Lambda-Carrageenan.
EINECSNumber 232-524-2
Recommended Storage Store at room temperature (15 to 30°C).
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.