Learn More
Enzo Life Sciences Irisin (human) polyclonal antibody (100µg)
SDP
Supplier:  Enzo Life Sciences ENZABS2560100
Encompass_Preferred
Immunogen: Recombinant human Irisin (active). Source: Purified from rabbit serum. Species reactivity: Human. Applications: ELISA, IP, WB. Purity: Protein A affinity purified and immunodepleted. Formulation: Liquid. In PBS containing 50% glycerol and 0.09% sodium azide.
Catalog No. 50-201-2801