Learn More
AdipoGen Irisin rec HEK293
SDP
Catalog No. 502562921
Encompass_Preferred
Recombinant protein. Irisin human, aa 32-143 /mouse, aa 29-140 is FLAG -tagged at the N-terminus. Irisin has 100% identity between mouse, rat, monkey and human. Source HEK 293 cells. Lyophilized. Contains PBS. Irisin is a recently described exercise-induced hormone secreted by skeletal muscle in mice and humans. Irisin activates beige fat cells beige cells have a gene expression pattern distinct from either white or brown fat and are preferentially sensitive to the polypeptide hormone Irisin. Irisin is cleaved from the type I membrane protein FNDC5 and improves systemic metabolism by increasing energy expenditure.
Catalog No. 50-256-2921 Supplier AdipoGen Supplier No. AG40B0102C010
Recombinant protein. Irisin human, aa 32-143 /mouse, aa 29-140 is FLAG -tagged at the N-terminus. Irisin has 100% identity between mouse, rat, monkey and human. Source HEK 293 cells. Lyophilized. Contains PBS. Irisin is a recently described exercise-induced hormone secreted by skeletal muscle in mice and humans. Irisin activates beige fat cells beige cells have a gene expression pattern distinct from either white or brown fat and are preferentially sensitive to the polypeptide hormone Irisin. Irisin is cleaved from the type I membrane protein FNDC5 and improves systemic metabolism by increasing energy expenditure.