Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Iristectorin A | 37744-61-9 | 1 ML
SDP

Catalog No. 5000419121
Encompass_Preferred

Iristectorin A is a natural product isolated from *Iris tectorum*. It is classified under Flavonoids, Isoflavones, Phenols, and Polyphenols. It exhibits anti-cancer activities, specifically noted in breast cancer. It is intended for research use only.

  • Molecular weight: 492.43
  • Formula: C23H24O12
  • Purity: 99.24%
  • Appearance: Solid
  • Color: Off-white to light yellow
  • Solubility: Soluble in DMSO at 100 mg/mL (203.07 mM).

Catalog No. 50-004-19121 Supplier Medchemexpress LLC Supplier No. HYN682010MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Iristectorin A is a natural product isolated from *Iris tectorum*. It is classified under Flavonoids, Isoflavones, Phenols, and Polyphenols. It exhibits anti-cancer activities, specifically noted in breast cancer. It is intended for research use only.

  • Molecular weight: 492.43
  • Formula: C23H24O12
  • Purity: 99.24%
  • Appearance: Solid
  • Color: Off-white to light yellow
  • Solubility: Soluble in DMSO at 100 mg/mL (203.07 mM).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.