Learn More
Medchemexpress LLC IRL-1620 | 142569-99-1 | 99.7% | 1820.95 | 1 MG
SDP
Catalog No. 5000391632
Encompass_Preferred
IRL-1620 is a potent and selective endothelin receptor type B (ETB) agonist. It is the most potent and specific ligand for the ETB receptor, with a Ki of 16 pM, showing 60 times more selectivity for the ETB receptor than ET-3. This compound is for research use only.
  • Induces contractions of the guinea pig trachea
  • Increases cytosolic Ca2+ in the vascular endothelium
  • Relaxes norepinephrine-stimulated tone in rat aorta
  • Improves acquisition and retention in the water maze task
  • Enhances angiogenic and neurogenic remodeling
  • Reduces cognitive impairment induced by Aβ in rats
  • Enhances the number of blood vessels labeled with VEGF
Catalog No. 50-003-91632 Supplier Medchemexpress LLC Supplier No. HY164651MG
IRL-1620 is a potent and selective endothelin receptor type B (ETB) agonist. It is the most potent and specific ligand for the ETB receptor, with a Ki of 16 pM, showing 60 times more selectivity for the ETB receptor than ET-3. This compound is for research use only.
  • Induces contractions of the guinea pig trachea
  • Increases cytosolic Ca2+ in the vascular endothelium
  • Relaxes norepinephrine-stimulated tone in rat aorta
  • Improves acquisition and retention in the water maze task
  • Enhances angiogenic and neurogenic remodeling
  • Reduces cognitive impairment induced by Aβ in rats
  • Enhances the number of blood vessels labeled with VEGF