Learn More
Medchemexpress LLC HY-103460 5mg Medchemexpress, IRL 2500 CAS:169545-27-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1034605MG
Encompass_Preferred
Medchemexpress, HY-103460 5mg IRL 2500 CAS:169545-27-1 IRL 2500 is a potent Endothelin receptor antagonist. IRL 2500 shows IC50 values of 1.3 and 94 nM for ETB and ETA receptors, respectively. IRL 2500 inhibits ETB receptor-mediated blood pressure increase and renal vascular resistance in rats in vivo. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8113