Learn More
R E Carroll Inc IRM 901 Oil 1 Gallon
SDP
Catalog No. NC0703778
Encompass
Testing oil for ASTM D471 Standard Test Method to evaluate the comparative ability of rubber and rubber-like compositions to withstand the effect of liquids.
Catalog No. NC0703778 Supplier R E Carroll Inc Supplier No. IRM901
Testing oil for ASTM D471 Standard Test Method to evaluate the comparative ability of rubber and rubber-like compositions to withstand the effect of liquids.