Learn More
R E Carroll Inc IRM 902 Oil 1 Gal
SDP
Catalog No. NC0376235
Encompass
Testing oil for ASTM D471 Standard Test Method to evaluate the comparative ability of rubber and rubber-like compositions to withstand the effect of liquids.
Catalog No. NC0376235 Supplier R E Carroll Inc Supplier No. IRM902
Testing oil for ASTM D471 Standard Test Method to evaluate the comparative ability of rubber and rubber-like compositions to withstand the effect of liquids.