Learn More
AdipoGen Iromycin A
SDP
Catalog No. 502562028
Encompass_Preferred
Chemical. CAS 213137-53-2. Formula C19H29NO2. MW 303.4. Isolated from Streptomyces bottropensis Gö-Dra17. Pyridone metabolite. Nitric oxide synthase NOS inhibitor, showing selectivity for eNOS NOS III versus nNOS NOS I. Thaxtomine biosynthesis inhibitor. Mitochondrial electron transport chain inhibitor.
Catalog No. 50-256-2028 Supplier AdipoGen Supplier No. BVT0262M001
Chemical. CAS 213137-53-2. Formula C19H29NO2. MW 303.4. Isolated from Streptomyces bottropensis Gö-Dra17. Pyridone metabolite. Nitric oxide synthase NOS inhibitor, showing selectivity for eNOS NOS III versus nNOS NOS I. Thaxtomine biosynthesis inhibitor. Mitochondrial electron transport chain inhibitor.