Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iron(0) pentacarbonyl, AcroSeal™

Catalog No. AC187621000
Change view
Click to view available options
100mL; AcroSeal glass bottle packed in tin can
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC187621000 100mL; AcroSeal glass bottle packed in tin can
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AC187621000 Supplier Thermo Scientific Chemicals Supplier No. 187621000
Only null left
Add to Cart
This item is not returnable. View return policy

CAS: 13463-40-6 | C5FeO5 | 195.90 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Boiling Point 103.0°C
Melting Point -20.0°C
Color Brown to Orange-Yellow
CAS 13463-40-6
Physical Form Liquid
Quantity 100 mL
Molecular Formula C5FeO5
Linear Formula Fe(CO)5
MDL Number MFCD00011001
UN Number 1994
Flash Point −15°C
Merck Index 15,5143
Synonym Pentacarbonyl iron
Solubility Information Solubility in water: insoluble
InChI Key FYOFOKCECDGJBF-UHFFFAOYSA-N
SMILES [Fe].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+]
IUPAC Name pentakis(methanidylidyneoxidanium) iron
Molecular Weight (g/mol) 195.90
Formula Weight 195.9
Infrared Spectrum Authentic
Packaging AcroSeal™ Glass bottle
Specific Gravity 1.49
Density 1.4900g/mL
Chemical Name or Material Iron(0) pentacarbonyl
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Fatal in contact with skin.
Fatal if inhaled.
Highly flammable liquid and vapour.
Fatal if swallowed.
Causes damage to organs through prolonged or repeated exposure if inhaled.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/
EINECSNumber 236-670-8

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.