Learn More
Aqua Solutions Iron Std. 10.0 mg/L 1 ml = 0.01 mg Fe ASTM E-200,E-394-15 (250ML)
SDP
Catalog No. NC0095486
Encompass_Preferred
Iron Std. 10.0 mg/L 1 ml = 0.01 mg Fe ASTM E-200,E-394-15 (250ML)
Catalog No. NC0095486 Supplier Aqua Solutions Supplier No. 4851250ML
Add to Cart
Add to Cart
Iron Std. 10.0 mg/L 1 ml = 0.01 mg Fe ASTM E-200,E-394-15 (250ML)