Learn More
Strem, An Ascensus Company CAS# 7439-89-6. 250g. Iron chips (99.95%). MFCD00010999
SDP
Catalog No. 5090114687
Encompass
CAS# 7439-89-6. 250g. Iron chips (99.95%). MFCD00010999. Molecular Weight: 55.80. Molecular Formula: Fe. Color/form: <2mm thick x <2mm x <25mm Strem# 93-2664. http://www.strem.com/catalog/v/93-2664/
Catalog No. 50-901-14687 Supplier Strem, An Ascensus Company Supplier No. 932664250G
CAS# 7439-89-6. 250g. Iron chips (99.95%). MFCD00010999. Molecular Weight: 55.80. Molecular Formula: Fe. Color/form: <2mm thick x <2mm x <25mm Strem# 93-2664. http://www.strem.com/catalog/v/93-2664/