Learn More
TARGETMOL CHEMICALS INC iron chloride hexahydrate 25MG
SDP
Catalog No. 502880217
Encompass_Preferred
Also available in 1 mL, 5 mg, 10 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. iron chloride hexahydrate promoted selective hydroxylation and chlorination of benzyl ketone derivatives for the construction of hetero-quaternary scaffolds.
Catalog No. 50-288-0217 Supplier TARGETMOL CHEMICALS INC Supplier No. T962425MG
Also available in 1 mL, 5 mg, 10 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. iron chloride hexahydrate promoted selective hydroxylation and chlorination of benzyl ketone derivatives for the construction of hetero-quaternary scaffolds.