Learn More
AdipoGen Iron Colorimetric Assay Kit (Ferrozine Method)
SDP
Supplier:  AdipoGen JAICFE005
Encompass_Preferred
Iron Colorimetric Assay Kit (Ferrozine Method), AdipoGen Life Sciences. This MC-Reagent Iron Assay Kit is a direct colorimetric assay based on the FerroZine¿ method without deproteinization of the sample. A weak acid buffer dissociates iron from the transferrin-iron complex and is reduced by a reductant (Ferric >> Ferrous). Ferrous ions form a complex with the chromogen Ferrozine. The color intensity is proportional to the iron concentration in the sample. Absorbance of the Fe2+-complex is measured at 560nm. Wavelength range of sensitivity: 540 ~ 580nm. Features of this Assay: Quick & Easy to use ¿ Species independent ¿ Highly sensitive, stable and suitable for high-throughput testing ¿ No toxic substances ¿ For cell lysates, serum, plasma and wide variety of biological samples
Catalog No. 50-101-5632