Learn More
Strem, An Ascensus Company CAS 635680-56-7. 1g. Bis(NN-di-t-butylacetamidinato)iron (II) min. 98%. MFCD08705281
SDP
Supplier:  Strem, An Ascensus Company 2601451G
Encompass
CAS 635680-56-7. 1g. Bis(NN-di-t-butylacetamidinato)iron (II) min. 98%. MFCD08705281. Molecular Weight 394.42. Molecular Formula C20H42N4Fe. Color/form dark gray solid Strem 26-0145. http//www.strem.com/catalog/v/26-0145/
Catalog No. 50-211-0786