Learn More
Strem, An Ascensus Company CAS# 12068-85-8. 1kg. Iron disulfide (Iron pyrite), 95%. MFCD00064690
SDP
Catalog No. 5090111098
Encompass
CAS# 12068-85-8. 1kg. Iron disulfide (Iron pyrite), 95%. MFCD00064690. Molecular Weight: 119.97. Molecular Formula: FeS2. Color/form: -20 mesh pwdr. Strem# 26-2400. http://www.strem.com/catalog/v/26-2400/
Catalog No. 50-901-11098 Supplier Strem, An Ascensus Company Supplier No. 2624001000G
CAS# 12068-85-8. 1kg. Iron disulfide (Iron pyrite), 95%. MFCD00064690. Molecular Weight: 119.97. Molecular Formula: FeS2. Color/form: -20 mesh pwdr. Strem# 26-2400. http://www.strem.com/catalog/v/26-2400/