Learn More
Strem, An Ascensus Company CAS# 17685-52-8. 10g. Iron dodecacarbonyl (Stabilized with 5-10% methanol). MFCD00135617
SDP
Catalog No. 5090111100
Encompass
CAS# 17685-52-8. 10g. Iron dodecacarbonyl (Stabilized with 5-10% methanol). MFCD00135617. Molecular Weight: 503.67. Molecular Formula: Fe3(CO)12. Color/form: black xtl. Strem# 26-2500. http://www.strem.com/catalog/v/26-2500/
Catalog No. 50-901-11100 Supplier Strem, An Ascensus Company Supplier No. 26250010G
CAS# 17685-52-8. 10g. Iron dodecacarbonyl (Stabilized with 5-10% methanol). MFCD00135617. Molecular Weight: 503.67. Molecular Formula: Fe3(CO)12. Color/form: black xtl. Strem# 26-2500. http://www.strem.com/catalog/v/26-2500/