Learn More
Sigma Aldrich Fine Chemicals Biosciences Iron Fe standard solution
Catalog No. 502908248
Encompass
This Standard Reference Material (SRM) is intended for use as a primary calibration standard for the quantitative determination of iron. A unit of SRM 3126a consists of five 10 mL sealed borosilicate glass ampoules of an acidified aqueous SOLUTIONon prepared gravimetrically to contain a known mass fraction of iron. The SOLUTIONon contains nitric acid at a volume fraction of approximately 10 % equivalent to an amount-of-substance concentration (molarity) of approximately 1.6 mol/L. For more information please refer to the COA and SDS.SRM 3126a cert SRM 3126a SDS
Catalog No. 50-290-8248 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. NIST3126A
This Standard Reference Material (SRM) is intended for use as a primary calibration standard for the quantitative determination of iron. A unit of SRM 3126a consists of five 10 mL sealed borosilicate glass ampoules of an acidified aqueous SOLUTIONon prepared gravimetrically to contain a known mass fraction of iron. The SOLUTIONon contains nitric acid at a volume fraction of approximately 10 % equivalent to an amount-of-substance concentration (molarity) of approximately 1.6 mol/L. For more information please refer to the COA and SDS.SRM 3126a cert SRM 3126a SDS