Learn More
Perkin Elmer US LLC Iron (Fe) Atomax Hollow Cathode Lamp
Supplier:  Perkin Elmer US LLC N2025317
Encompass_Preferred
Single Element Atomax™ Hollow Cathode Lamp (HCL) for the detection of elemental Iron (Fe). PerkinElmer Atomax™ 1.5 in. Hollow Cathode Lamps are crafted to the same high standards that have made our 2 in. Lumina™ Hollow Cathode Lamps the best in the industry. Drawing on more than 40 years of HCL design and manufacturing expertise, Atomax™ lamps now offer laboratories around the world a dependable, top-quality lighting source, regardless of the brand of AA instruments used. All 1.5 in. Atomax™ Hollow Cathode Lamps are thoroughly tested to ensure maximum light output and spectral purity. Comparative results show that Atomax lamps provide equal or better performance than other AA lamps in the market. Precision engineered for high sensitivity, stable light output, low noise, and long life, you can rely on Atomax™ lamps for the accurate results you need.
Catalog No. 50-906-337