Learn More
Iron HR Reagent Set, Lovibond™
Catalog No. 09050145 Shop All Tintometer Group Products
Change view
Click to view available options
Volume (Metric):
50 mL
Catalog No. Volume (Metric)
09-050-145 50 mL
1 options
Catalog No. 09-050-145 Supplier Tintometer Group Supplier No. 56R023590
Add to Cart
Add to Cart
Comes with Iron reagent FE6 and Hardness Total Buffer TH2, for use with Lovibond method M227.
  • Test range: 0.1 to 10 mg/L
  • Must be able to boil sample for 20 minutes
Specifications
pH Range 0 to 14
Volume (Metric) 50 mL
Type Iron Reagent Set