Learn More
Iron ICP Standard, Ricca Chemical
SDP
Catalog No. 42014 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
120 mL
Catalog No. Quantity
4201-4 120 mL
1 options
Catalog No. 4201-4 Supplier Ricca Chemical Company Supplier No. 42014
1mL = 1mg Fe, 1000ppm Fe. (Fe in 5% HNO3) (Class: 8, UN3264, Pkg Grp: III, Corrosive Liquid, Acidic, Inorganic, n.o.s, Contains: Nitric Acid)
Specifications
Quantity 120 mL