Learn More
Crescent Chemical Co Inc IRON ICP STD 100ML
SDP
Catalog No. 5072013
Encompass_Preferred
Iron, HN03 Single Element Plasma Emission Spectroscopy Standards 1,000 UgmL NIST Traceable CAS Size - 100mL Catalog Number - 8552
Catalog No. 50-720-13 Supplier Crescent Chemical Co Inc Supplier No. IS8552 (100ML)
Iron, HN03 Single Element Plasma Emission Spectroscopy Standards 1,000 UgmL NIST Traceable CAS Size - 100mL Catalog Number - 8552