Learn More
SPEX CERTIPREP LLC 3P IRON ICP STD 100ML 1000PPM
Catalog No. NC0216589
Encompass_Preferred
(HPS by Zeptometrix) Fe 1000ug/mL in 2% HCl Exp Date: 18 Months This standard is traceable to NIST and certified Guide 34 and 17025.
Catalog No. NC0216589 Supplier SPEX CERTIPREP LLC 3P Supplier No. 1000262100
(HPS by Zeptometrix) Fe 1000ug/mL in 2% HCl Exp Date: 18 Months This standard is traceable to NIST and certified Guide 34 and 17025.