Learn More
Strem, An Ascensus Company CAS# 3094-87-9. 5g. Iron(II) acetate, anhydrous, 97%. MFCD00058909
SDP
Catalog No. 5090114701
Encompass
CAS# 3094-87-9. 5g. Iron(II) acetate, anhydrous, 97%. MFCD00058909. Molecular Weight: 173.94. Molecular Formula: Fe(OOCCH3)2. Color/form: off-white to light brown pwdr. Strem# 93-2678. http://www.strem.com/catalog/v/93-2678/
Catalog No. 50-901-14701 Supplier Strem, An Ascensus Company Supplier No. 9326785G
CAS# 3094-87-9. 5g. Iron(II) acetate, anhydrous, 97%. MFCD00058909. Molecular Weight: 173.94. Molecular Formula: Fe(OOCCH3)2. Color/form: off-white to light brown pwdr. Strem# 93-2678. http://www.strem.com/catalog/v/93-2678/