Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iron(II) acetate, anhydrous, Fe 29.5% min

Catalog No. AA3114022
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3114022 100 g
AA3114014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3114022 Supplier Thermo Scientific Chemicals Supplier No. 03114022
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 3094-87-9 | C4H6FeO4 | 173.93 g/mol

Iron(II) acetate is used as a mordant by the dye industry, as a catalyst in organic oxidation reactions and in carbon nanotube synthesis. It is used as a precursor in the production of other iron compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3094-87-9
Molecular Formula C4H6FeO4
Molecular Weight (g/mol) 173.93
MDL Number MFCD00058909
InChI Key LNOZJRCUHSPCDZ-UHFFFAOYSA-L
Synonym iron ii acetate, ferrous acetate, iron diacetate, iron di acetate, iron 2+ acetate, acetic acid, iron 2+ salt, acetic acid, iron 2+ salt 2:1, unii-l80i7m6d3q, ferrousacetate
PubChem CID 18344
SMILES [Fe++].CC([O-])=O.CC([O-])=O

Specifications

Melting Point 190°C to 200°C (decomposition)
Color Brown
Physical Form Lump/Powder
Quantity 100 g
Odor Characteristic
Sensitivity Air and moisture sensitive
Solubility Information Soluble in water.
IUPAC Name λ²-iron(2+) diacetate
Formula Weight 173.94
Chemical Name or Material Iron(II) acetate, anhydrous
Hazard Category H315-H319-H335-H500
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 221-441-7
RTECSNumber AI3850000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.