Learn More
Strem, An Ascensus Company CAS# 7789-46-0. 10g. Iron(II) bromide, anhydrous, 98%. MFCD00016081
SDP
Catalog No. 5090114654
Encompass
CAS# 7789-46-0. 10g. Iron(II) bromide, anhydrous, 98%. MFCD00016081. Molecular Weight: 215.67. Molecular Formula: FeBr2. Color/form: orange pwdr. Strem# 93-2629. http://www.strem.com/catalog/v/93-2629/
Catalog No. 50-901-14654 Supplier Strem, An Ascensus Company Supplier No. 93262910G
CAS# 7789-46-0. 10g. Iron(II) bromide, anhydrous, 98%. MFCD00016081. Molecular Weight: 215.67. Molecular Formula: FeBr2. Color/form: orange pwdr. Strem# 93-2629. http://www.strem.com/catalog/v/93-2629/