Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Iron(II) chloride, 97%, anhydrous, Acros Organics
Click to view available options
Quantity:
25g
250g
Packaging:
Glass bottle
Specifications
Assay Percent Range | 97% |
Linear Formula | FeCl2 |
Merck Index | 15,407 |
Solubility Information | Solubility in water: 644g/L |
Formula Weight | 126.75 |
Physical Form | Powder |
Percent Purity | 97% |
Packaging | Glass bottle |
Solubility | Solubility in water: 644g/L. |
Boiling Point | 1026.0°C |
Show More |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with
GHS Signal Word: Danger
EINECSNumber : 231-843-4