Learn More
Strem, An Ascensus Company CAS# 7758-94-3. 25g. Iron(II) chloride, anhydrous, 98%. MFCD00011004
SDP
Catalog No. 5090114656
Encompass
CAS# 7758-94-3. 25g. Iron(II) chloride, anhydrous, 98%. MFCD00011004. Molecular Weight: 126.75. Molecular Formula: FeCl2. Color/form: tan pwdr. Strem# 93-2631. http://www.strem.com/catalog/v/93-2631/
Catalog No. 50-901-14656 Supplier Strem, An Ascensus Company Supplier No. 93263125G
CAS# 7758-94-3. 25g. Iron(II) chloride, anhydrous, 98%. MFCD00011004. Molecular Weight: 126.75. Molecular Formula: FeCl2. Color/form: tan pwdr. Strem# 93-2631. http://www.strem.com/catalog/v/93-2631/