Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iron(II) chloride, anhydrous, 99.5% (metals basis)

Catalog No. aa3114106
Change view
Click to view available options
Quantity:
5 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3114106 5 g
AA3114118 50 g
AA3114130 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA3114106 Supplier Thermo Scientific Chemicals Supplier No. 03114106
Only null left
Add to Cart

CAS: 7758-94-3 | Cl2Fe | 126.75 g/mol

Iron(II) chloride is used in the synthesis of iron complexes like iron(II) hydroxide. It acts as a precursor to hydrated iron(III) oxides that are magnetic pigments. It serves as a reducing agent in organic synthesis. Also, used as a reducing flocculating agent in wastewater treatment, especially for wastes containing chromate. It is used as an intermediate, in pigments, processing aids and in solids separation agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7758-94-3
Molecular Formula Cl2Fe
Molecular Weight (g/mol) 126.75
MDL Number MFCD00011004
InChI Key NMCUIPGRVMDVDB-UHFFFAOYSA-L
IUPAC Name λ²-iron(2+) dichloride
SMILES [Cl-].[Cl-].[Fe++]

Specifications

Boiling Point 1023°C
Melting Point 674°C
pH 2.5
Physical Form Powder
Quantity 5 g
Linear Formula FeCl2
UN Number UN3260
Merck Index 14,4043
Solubility Information Freely soluble in water,alcohol,acetone;Soluble in water,alcohol and acetone; Slightly soluble in benzene; Insoluble in ether.
Formula Weight 126.75
Percent Purity 99.5%
Odor Odorless
Sensitivity Air sensitive; Hygroscopic
Density 3.160
Assay (metals basis)
Chemical Name or Material Iron(II) chloride, anhydrous
Show More Show Less
Hazard Category H302-H314-H335
Hazard Statement GHS H Statement
H314-H318-H302;
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.
EINECSNumber 231-843-4
RTECSNumber NO5400000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.