Learn More
Strem, An Ascensus Company CAS# 13478-10-9. 1kg. Iron(II) chloride tetrahydrate, 99%. MFCD00149709
SDP
Catalog No. 5090114659
Encompass
CAS# 13478-10-9. 1kg. Iron(II) chloride tetrahydrate, 99%. MFCD00149709. Molecular Weight: 126.75 (198.81). Molecular Formula: FeCl2∙4H2O. Color/form: light green xtl. Strem# 93-2632. http://www.strem.com/catalog/v/93-2632/
Catalog No. 50-901-14659 Supplier Strem, An Ascensus Company Supplier No. 9326321000G
CAS# 13478-10-9. 1kg. Iron(II) chloride tetrahydrate, 99%. MFCD00149709. Molecular Weight: 126.75 (198.81). Molecular Formula: FeCl2∙4H2O. Color/form: light green xtl. Strem# 93-2632. http://www.strem.com/catalog/v/93-2632/