Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Iron(II) chloride tetrahydrate

Catalog No. 3800245G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
38-002-45G 5 g
38-002-425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 38-002-45G Supplier Sigma Aldrich Supplier No. 3800245G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Iron(II) chloride tetrahydrate is a hydrated metallic halide. Crystal structure studies suggest that it crystallizes in monoclinic system having space group P21/c.Iron(II) chloride tetrahydrate (Ferrous chloride tetrahydrate, FeCl2-+4H2O) has been used in the preparation of bentonite/iron nanoparticles.It may be used in the following studies:Preparation of composites of multiwall carbon nanotubes (MWCNTs)/nano-iron oxide. Preparation of magnetite (Fe3O4) nanoparticles. Solid-phase synthesis of [Fe(phen)3]Cl2-+nH2O (phen = 1,10-phenanthroline).4

Specifications

CAS 13478-10-9
Molecular Formula Cl2Fe -+ 4H2O
Linear Formula FeCl2 -+ 4H2O
MDL Number MFCD00149709
Quantity 5 g
Synonym Ferrous chloride tetrahydrate
Molecular Weight (g/mol) 198.81
Percent Purity 99.99% trace metals basis
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.