Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iron(Ii) Chloride Tetrahydrate, Solstice

Catalog No. 6000827 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
1 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-008-27 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-008-27 Supplier Solstice Supplier No. 2202991KG

missing translation for 'validMainframeMsg'

Reagent Grade, 98%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13478-10-9
Molecular Formula Cl2FeH8O4
Molecular Weight (g/mol) 198.805
MDL Number MFCD00149709
InChI Key WSSMOXHYUFMBLS-UHFFFAOYSA-L
Synonym ferrous chloride tetrahydrate, iron ii chloride tetrahydrate, iron dichloride tetrahydrate, cl2fe.4h2o, fecl2.4h2o, iron chloride, tetrahydrate, dichloroiron-water 1/4, iron ii chloride tetra hydrate, ae>> nciu eae r masculineiii
PubChem CID 16211588
ChEBI CHEBI:86249
IUPAC Name dichloroiron;tetrahydrate
SMILES O.O.O.O.Cl[Fe]Cl

Specifications

Quantity 1 kg
Linear Formula FeCl2 · 4H2O
UN Number 3260
Formula Weight 198.81g/mol
Percent Purity 98%
Grade ReagentPlus
Vapor Pressure 10mmHg (693°C)
Chemical Name or Material Iron(Ii) Chloride Tetrahydrate
Hazard Statement P280-P305 + P351 + P338-P310
Precautionary Statement H302-H314
EINECSNumber 231-843-4
RTECSNumber NO5600000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.