Learn More
Strem, An Ascensus Company CAS# 7789-28-8. 25g. Iron(II) fluoride, anhydrous, 99%. MFCD00016093
SDP
Catalog No. 5090114700
Encompass
CAS# 7789-28-8. 25g. Iron(II) fluoride, anhydrous, 99%. MFCD00016093. Molecular Weight: 93.84. Molecular Formula: FeF2. Color/form: -80 mesh off-white pwdr. Strem# 93-2675. http://www.strem.com/catalog/v/93-2675/
Catalog No. 50-901-14700 Supplier Strem, An Ascensus Company Supplier No. 93267525G
CAS# 7789-28-8. 25g. Iron(II) fluoride, anhydrous, 99%. MFCD00016093. Molecular Weight: 93.84. Molecular Formula: FeF2. Color/form: -80 mesh off-white pwdr. Strem# 93-2675. http://www.strem.com/catalog/v/93-2675/