Learn More
Medchemexpress LLC Iron(II) fumarate (Ferrous fumarate) | 141-01-5 | 95.1% | 1 G
SDP
Catalog No. 5000413167
Encompass_Preferred
Iron (II) fumarate is an orally effective active dietary supplement. It can alleviate metabolic damage and damage to silkworms caused by cypermethrin and can be used for research on iron deficiency anemia.
  • In vitro: Can restore metabolic pathway disruption and enhance cell viability in BmN cells treated with cypermethrin.
  • In vivo: Alleviates damage caused by cypermethrin in silkworms.
  • In vivo: Induces oxidative stress in the liver and colon of rats.
  • In vivo: Alters intestinal morphology and microbiota in Yellow River carp, increasing diversity and richness.
Catalog No. 50-004-13167 Supplier Medchemexpress LLC Supplier No. HYB16511G
Iron (II) fumarate is an orally effective active dietary supplement. It can alleviate metabolic damage and damage to silkworms caused by cypermethrin and can be used for research on iron deficiency anemia.
  • In vitro: Can restore metabolic pathway disruption and enhance cell viability in BmN cells treated with cypermethrin.
  • In vivo: Alleviates damage caused by cypermethrin in silkworms.
  • In vivo: Induces oxidative stress in the liver and colon of rats.
  • In vivo: Alters intestinal morphology and microbiota in Yellow River carp, increasing diversity and richness.