Learn More
Medchemexpress LLC Iron(II) gluconate | 699014-53-4 | MFCD00150872 | 91.3% | 464.15 | C12H22FeO14·xH2O | 100 MG
SDP
Catalog No. 5000284496
Encompass_Preferred
Iron(II) gluconate is the ferrous salt of gluconic acid supplied as a light yellow to brown solid. It is used as a source of ferrous iron (Fe(II)) in biochemical assays and laboratory research; for research use only and not for human consumption.
  • Provides ferrous iron (Fe(II)) for biochemical and assay applications.
  • Assay/purity approximately 91.3%.
  • Available in small laboratory sizes (25 mg, 50 mg, 100 mg; larger sizes by quote).
  • Solid form with light yellow to brown appearance for ease of handling and storage.
  • Certificate of analysis, safety data sheet, and handling instructions available.
  • Intended for research use only; not for clinical or human use.
Catalog No. 50-002-84496 Supplier Medchemexpress LLC Supplier No. HYW698286100MG
Iron(II) gluconate is the ferrous salt of gluconic acid supplied as a light yellow to brown solid. It is used as a source of ferrous iron (Fe(II)) in biochemical assays and laboratory research; for research use only and not for human consumption.
  • Provides ferrous iron (Fe(II)) for biochemical and assay applications.
  • Assay/purity approximately 91.3%.
  • Available in small laboratory sizes (25 mg, 50 mg, 100 mg; larger sizes by quote).
  • Solid form with light yellow to brown appearance for ease of handling and storage.
  • Certificate of analysis, safety data sheet, and handling instructions available.
  • Intended for research use only; not for clinical or human use.