Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Iron(II) gluconate | 699014-53-4 | MFCD04037236 | 91.3% | 464.15 g·mol⁻¹ | C12H22FeO14·xH2O | 50 MG
SDP

Catalog No. 5000284811
Encompass_Preferred

Iron(II) gluconate is a biochemical reagent used in research on iron metabolism and iron-deficiency models. Supplied as a solid in milligram quantities for laboratory assays; for research use only, not for human use.

  • Suitable for biochemical assays and studies of iron metabolism.
  • Provided as a solid in small milligram quantities for laboratory use.
  • Assay approximately 91.3% (manufacturer reported).
  • Molecular weight ~464.15 g·mol⁻¹.
  • CAS number 699014-53-4; SDS and COA available.

Catalog No. 50-002-84811 Supplier Medchemexpress LLC Supplier No. HYW69828650MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Iron(II) gluconate is a biochemical reagent used in research on iron metabolism and iron-deficiency models. Supplied as a solid in milligram quantities for laboratory assays; for research use only, not for human use.

  • Suitable for biochemical assays and studies of iron metabolism.
  • Provided as a solid in small milligram quantities for laboratory use.
  • Assay approximately 91.3% (manufacturer reported).
  • Molecular weight ~464.15 g·mol⁻¹.
  • CAS number 699014-53-4; SDS and COA available.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.