Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iron(II,III) oxide, nanopowder, 97%

Catalog No. p-7052280
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4714114 25 g
AA4714122 100 g
AA4714136 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4714114 Supplier Thermo Scientific Chemicals Supplier No. 04714114
Only null left
Add to Cart

CAS: 1317-61-9 | Fe3O4 | 231.53 g/mol

Iron(II,III) oxide is mainly used as a black-pigment-catalyst in the Haber process and in the water gas shift reaction. It is used as a contrast-agent in MRI scanning and as a pigment in magnetic applications, polishing compounds, cosmetics, medicines, polymers, rubber, fillers, building, construction, appliances and magnetic inks.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1317-61-9
Molecular Formula Fe3O4
Molecular Weight (g/mol) 231.53
MDL Number MFCD00011010
InChI Key WTFXARWRTYJXII-UHFFFAOYSA-N
IUPAC Name diiron(3+) λ²-iron(2+) tetraoxidandiide
SMILES [O--].[O--].[O--].[O--].[Fe++].[Fe+3].[Fe+3]

Specifications

Melting Point 1538°C
Color Black
Physical Form Powder
Quantity 25 g
Merck Index 14,4040
Solubility Information Insoluble in water and organic solvents. Soluble in concentrated mineral acids.
Formula Weight 231.54
Percent Purity 97%
Odor Odorless
Refractive Index 3.0
Density 5.2 g/mL
Chemical Name or Material Iron(II,III) oxide
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 215-277-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.