Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Iron(II) Lactate Hydrate

Catalog No. 44953100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
44-953-100G 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 44-953-100G Supplier Sigma Aldrich Supplier No. 44953100G
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Reagent in preparation of carbon-based noble-metal-free PEFC cathode catalysts (oxygen reduction) for use in polymer electrolyte fuel cells (PEFCs)Used for formulation of chemoattractant coatings for artificial reef structures

Specifications

CAS 944797-51-7
Melting Point 242°C to 245°C
Molecular Formula C23H14BF7O
Linear Formula C23H14BF7O
Quantity 250 mg
Synonym Triphenylpyrylium photosensitizer; [T(p-F)PPT
Molecular Weight (g/mol) 450.16 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.