Learn More
Strem, An Ascensus Company CAS# 2980-59-8. 100g. Iron(II) stearate (9% Fe). MFCD00050715
SDP
Catalog No. 5090111107
Encompass
CAS# 2980-59-8. 100g. Iron(II) stearate (9% Fe). MFCD00050715. Molecular Weight: 622.81. Molecular Formula: Fe(O2C18H35)2. Color/form: orange pwdr. Strem# 26-2622. http://www.strem.com/catalog/v/26-2622/
Catalog No. 50-901-11107 Supplier Strem, An Ascensus Company Supplier No. 262622100G
CAS# 2980-59-8. 100g. Iron(II) stearate (9% Fe). MFCD00050715. Molecular Weight: 622.81. Molecular Formula: Fe(O2C18H35)2. Color/form: orange pwdr. Strem# 26-2622. http://www.strem.com/catalog/v/26-2622/