Learn More
Medchemexpress LLC Iron(II) sulfate heptahydrate | 7782-63-0 | MFCD00149172 | 99.5% | 278.01 | FeSO4·7H2O | 100g
SDP
Catalog No. 5000267522
Encompass_Preferred
Iron(II) sulfate heptahydrate for cell culture 99% (Ferrous sulfate heptahydrate for cell culture 99%) is an orally active iron salt Iron(II) sulfate heptahydrate for cell culture 99% replaces iron found in hemoglobin and myoglobin allowing for the transportation of oxygen via hemoglobin Iron(II) sulfate heptahydrate for cell culture 99% is mainly used for the prevention of iron-deficiency anemia Iron(II) sulfate heptahydrate for cell culture 99% also has anti-tumor effects on chronic myeloid leukemia and breast cancer[1][2][3]
Catalog No. 50-002-67522 Supplier Medchemexpress LLC Supplier No. HYY1103A100G
Iron(II) sulfate heptahydrate for cell culture 99% (Ferrous sulfate heptahydrate for cell culture 99%) is an orally active iron salt Iron(II) sulfate heptahydrate for cell culture 99% replaces iron found in hemoglobin and myoglobin allowing for the transportation of oxygen via hemoglobin Iron(II) sulfate heptahydrate for cell culture 99% is mainly used for the prevention of iron-deficiency anemia Iron(II) sulfate heptahydrate for cell culture 99% also has anti-tumor effects on chronic myeloid leukemia and breast cancer[1][2][3]