Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Iron(II) sulfate hydrate

Catalog No. 4502785G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45-027-85G 5 g
45-027-825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 45-027-85G Supplier Sigma Aldrich Supplier No. 4502785G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Iron(II) sulfate hydrate has been used to prepare ferrous ion solution to study the influence of iron ions on the thermal decomposition behaviour of hydroxylamine/water solutions.

Specifications

CAS 13463-43-9
Quantity 5 g
Molecular Formula FeO4S -+ xH2O
Linear Formula FeSO4 -+ xH2O
MDL Number MFCD00149722
Synonym Green vitriol; Iron vitriol; Sulfuric acid, iron(II) salt (1:1), hydrate
Molecular Weight (g/mol) 151.91
Percent Purity 99.999% trace metals basis
Density 1.9 g/mL (at 25°C)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.