Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iron(Ii) Sulfate Hydrate, Solstice

Catalog No. 6001933 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
50 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-019-33 50 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-019-33 Supplier Solstice Supplier No. 1235350KG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Puriss., meets analytical specification of BP, European Pharmacopoeia, exsiccated, 86.0-89.0% FeSO4 basis

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13463-43-9
Molecular Formula FeH2O5S
Molecular Weight (g/mol) 169.916
MDL Number MFCD00149722
InChI Key XBDUTCVQJHJTQZ-UHFFFAOYSA-L
Synonym ferrous sulfate monohydrate, iron 2+ sulfate monohydrate, ferrous sulfate hydrate, iron sulfate monohydrate, unii-rib00980vw, iron ii sulfate, monohydrate, iron ii sulfate hydrate, sulfuric acid, iron 2+ salt 1:1 , monohydrate, feroritard, feromax
PubChem CID 62712
ChEBI CHEBI:75834
IUPAC Name iron(2+);sulfate;hydrate
SMILES O.[O-]S(=O)(=O)[O-].[Fe+2]

Specifications

Physical Form Powder
Quantity 50 kg
Linear Formula FeSO4 · aq
Formula Weight 151.91g/mol (anhydrous basis)
Percent Purity 86.0 to 89.0%
Grade puriss.
Concentration or Composition (by Analyte or Components) 86 to 89.0% FeSO4 basis
Chemical Name or Material Iron(Ii) Sulfate Hydrate, Meets analytical specification of Ph. Eur.BP
Hazard Statement P273-P305 + P351 + P338-P501
Precautionary Statement H302-H315-H319-H410
EINECSNumber 231-753-5
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.