Learn More
Strem, An Ascensus Company CAS# 1317-37-9. 100g. Iron(II) sulfide (99%-Fe). MFCD00011013
SDP
Catalog No. 5090114670
Encompass
CAS# 1317-37-9. 100g. Iron(II) sulfide (99%-Fe). MFCD00011013. Molecular Weight: 87.91. Molecular Formula: FeS. Color/form: -60 + 200 mesh black pwdr. Strem# 93-2640. http://www.strem.com/catalog/v/93-2640/
Catalog No. 50-901-14670 Supplier Strem, An Ascensus Company Supplier No. 932640100G
CAS# 1317-37-9. 100g. Iron(II) sulfide (99%-Fe). MFCD00011013. Molecular Weight: 87.91. Molecular Formula: FeS. Color/form: -60 + 200 mesh black pwdr. Strem# 93-2640. http://www.strem.com/catalog/v/93-2640/