Learn More
Strem, An Ascensus Company CAS# 15283-51-9. 50g. Iron(II) tetrafluoroborate, 40-45% aqueous solution. MFCD00016079
SDP
Catalog No. 5090114660
Encompass
CAS# 15283-51-9. 50g. Iron(II) tetrafluoroborate, 40-45% aqueous solution. MFCD00016079. Molecular Weight: 229.45. Molecular Formula: Fe(BF4)2. Color/form: green liq. Strem# 93-2633. http://www.strem.com/catalog/v/93-2633/
Catalog No. 50-901-14660 Supplier Strem, An Ascensus Company Supplier No. 93263350G
CAS# 15283-51-9. 50g. Iron(II) tetrafluoroborate, 40-45% aqueous solution. MFCD00016079. Molecular Weight: 229.45. Molecular Formula: Fe(BF4)2. Color/form: green liq. Strem# 93-2633. http://www.strem.com/catalog/v/93-2633/