Learn More
Strem, An Ascensus Company Iron › Iron(III) acetylacetonate, 99%, 250 Grams
SDP
Catalog No. 5090111097
Encompass
Iron (III) acetylacetonate, 99%; Size: 250g; [14024-18-1]; Fe(CH3COCHCOCH3)3; Mol Wt:353.18; Color & Form: orange to red xtl.
Catalog No. 50-901-11097 Supplier Strem, An Ascensus Company Supplier No. 262300250G
Iron (III) acetylacetonate, 99%; Size: 250g; [14024-18-1]; Fe(CH3COCHCOCH3)3; Mol Wt:353.18; Color & Form: orange to red xtl.