Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iron(III) acrylate, 96%, Thermo Scientific™

Catalog No. AA4200122
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4200122 100 g
AA4200114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4200122 Supplier Thermo Scientific Chemicals Supplier No. 4200122

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 55488-18-1
Molecular Formula C9H9FeO6
Molecular Weight (g/mol) 269.01
MDL Number MFCD00080380
InChI Key WNWBIDPJHFYYLM-UHFFFAOYSA-K
Synonym iron iii acrylate, feracryl, pk 2 acrylic polymer, poly acrylic acid iron salt, iron acrylate, 2-propenoic acid, homopolymer, iron salt, iron triacrylate, 79-10-7 parent, iron 3+ acrylate, 2-propenoic acid, iron 3+ salt
PubChem CID 171450
IUPAC Name iron(3+);prop-2-enoate
SMILES C=CC(=O)[O-].C=CC(=O)[O-].C=CC(=O)[O-].[Fe+3]

Specifications

Melting Point 190°C to 200°C (decomposition)
Quantity 100 g
Sensitivity Air and moisture sensitive
Formula Weight 269.01
Percent Purity 96%
Physical Form Powder
Chemical Name or Material Iron(III) acrylate
Hazard Category H317
Hazard Statement GHS H Statement
H317
May cause an allergic skin reaction.
Precautionary Statement P261-P272-P280g-P302+P352-P333+P313-P363-P501c
EINECSNumber 259-667-3
TSCA Yes
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.