Learn More
Strem, An Ascensus Company CAS# 10031-26-2. 50G. Iron(III) bromide, anhydrous, 99%. MFCD00016082
SDP
Catalog No. 501963556
Encompass
CAS# 10031-26-2. 50G. Iron(III) bromide, anhydrous, 99%. MFCD00016082. Molecular Formula: FeBr3. Molecular Weight: 295.57. Color/Form: reddish-black xtl. Stability: hygroscopic. Strem# 93-2665. www.strem.com/catalog/v3/93-2665/
Catalog No. 50-196-3556 Supplier Strem, An Ascensus Company Supplier No. 93266550G
CAS# 10031-26-2. 50G. Iron(III) bromide, anhydrous, 99%. MFCD00016082. Molecular Formula: FeBr3. Molecular Weight: 295.57. Color/Form: reddish-black xtl. Stability: hygroscopic. Strem# 93-2665. www.strem.com/catalog/v3/93-2665/