Learn More
Strem, An Ascensus Company CAS# 7705-08-0. 5kg. Iron(III) chloride, anhydrous, 98%. MFCD00011005
SDP
Catalog No. 5090114633
Encompass
CAS# 7705-08-0. 5kg. Iron(III) chloride, anhydrous, 98%. MFCD00011005. Molecular Weight: 162.22. Molecular Formula: FeCl3. Color/form: black pwdr. Strem# 93-2607. http://www.strem.com/catalog/v/93-2607/
Catalog No. 50-901-14633 Supplier Strem, An Ascensus Company Supplier No. 9326075000G
CAS# 7705-08-0. 5kg. Iron(III) chloride, anhydrous, 98%. MFCD00011005. Molecular Weight: 162.22. Molecular Formula: FeCl3. Color/form: black pwdr. Strem# 93-2607. http://www.strem.com/catalog/v/93-2607/