Learn More
Strem, An Ascensus Company CAS# 10025-77-1. 250g. Iron(III) chloride hexahydrate, 97+% (ACS). MFCD00149712
SDP
Catalog No. 5090114630
Encompass
CAS# 10025-77-1. 250g. Iron(III) chloride hexahydrate, 97+% (ACS). MFCD00149712. Molecular Weight: 162.22 (270.32). Molecular Formula: FeCl3∙6H2O. Color/form: yellow lump Strem# 93-2606. http://www.strem.com/catalog/v/93-2606/
Catalog No. 50-901-14630 Supplier Strem, An Ascensus Company Supplier No. 932606250G
CAS# 10025-77-1. 250g. Iron(III) chloride hexahydrate, 97+% (ACS). MFCD00149712. Molecular Weight: 162.22 (270.32). Molecular Formula: FeCl3∙6H2O. Color/form: yellow lump Strem# 93-2606. http://www.strem.com/catalog/v/93-2606/