Learn More
Sigma Aldrich Fine Chemicals Biosciences Iron(III) chloride hexahydrate, 10025-77-1, MFCD00149712, 250g
Catalog No. NC1365928
Encompass
Linear Formula FeCl3 · 6H2O, ≥98%, Molecular Weight 270.30, bp 280-285 °C (lit.), mp 37 °C (lit.), reagent grade, chunks, Synonym: Ferric chloride hexahydrate
Catalog No. NC1365928 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. F2877250G
Linear Formula FeCl3 · 6H2O, ≥98%, Molecular Weight 270.30, bp 280-285 °C (lit.), mp 37 °C (lit.), reagent grade, chunks, Synonym: Ferric chloride hexahydrate